Anti-SUMO1 Rabbit polyclonal antibody

ABCAAB3819-50EA 744 CHF
ABCAAB3819-50
Anti-SUMO1 Rabbit polyclonal antibody
Type: Primary
Antigen: SUMO1 (small ubiquitin-like modifier 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR