Anti-SNF2H Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB3749-100EA 486.49 CHF
ABCAAB3749-100
Anti-SNF2H Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNF2H
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse