Anti-Sumo 2+3 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3742-100EA 513.55 CHF
ABCAAB3742-100
Anti-Sumo 2+3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Sumo 2+3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR