Anti-BRD2 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB3718-50EA 933 CHF
ABCAAB3718-50
Anti-BRD2 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BRD2 (bromodomain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR