Anti-ING4 Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB3714-100EA 933 CHF
ABCAAB3714-100
Anti-ING4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ING4 (inhibitor of growth family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR