Anti-SOCS4 Rabbit polyclonal antibody

ABCAAB3694-50EA 737 CHF
ABCAAB3694-50
Anti-SOCS4 Rabbit polyclonal antibody
Type: Primary
Antigen: SOCS4 (suppressor of cytokine signaling 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR