Anti-SOCS3 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3693-50EA 945 CHF
ABCAAB3693-50
Anti-SOCS3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOCS3 (suppressor of cytokine signaling 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR