Anti-SOCS1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3691-50EA 933 CHF
ABCAAB3691-50
Anti-SOCS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR