Anti-FEM1B Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB3686-100EA 870 CHF
ABCAAB3686-100
Anti-FEM1B Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FEM1B (fem-1 homolog b (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR