Anti-MRE11A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3621-50EA 933 CHF
ABCAAB3621-50
Anti-MRE11A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MRE11A (MRE11 meiotic recombination 11 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR