Anti-RAD17 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB3620-50EA 534.19 CHF
ABCAAB3620-50
Anti-RAD17 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD17 (RAD17 homolog (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR