Anti-NDC80 Mouse Monoclonal Antibody [clone: 9G3]

Supplier: Abcam
ABCAAB3613-100EA 456.49 CHF
ABCAAB3613-100
Anti-NDC80 Mouse Monoclonal Antibody [clone: 9G3]
Antibodies
Type: Primary
Antigen: NDC80 (NDC80 kinetochore complex component)
Clonality: monoclonal
Clone: 9G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR