Anti-Proprotein Convertase 2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3533-100EA 477.24 CHF
ABCAAB3533-100
Anti-Proprotein Convertase 2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Proprotein Convertase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR