Anti-G Protein alpha Inhibitor 1+2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB3522-100EA 524.65 CHF
ABCAAB3522-100
Anti-G Protein alpha Inhibitor 1+2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: G Protein alpha Inhibitor 1+2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR