Anti-Serotonin N-acetyltransferase Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3505-50EA 508.36 CHF
ABCAAB3505-50
Anti-Serotonin N-acetyltransferase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Serotonin N-acetyltransferase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR