Anti-BRIP1 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB3502-100EA 920 CHF
ABCAAB3502-100
Anti-BRIP1 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: BRIP1 (BRCA1 interacting protein C-terminal helicase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR