Anti-PPARA Rabbit polyclonal antibody

ABCAAB3485-100EA 660 CHF
ABCAAB3485-100
Anti-PPARA Rabbit polyclonal antibody
Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR