Anti-MYO7A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3481-200EA 518.74 CHF
ABCAAB3481-200
Anti-MYO7A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MYO7A (myosin VIIA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR