Anti-alpha 1 Adrenergic Receptor Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB3462-100EA 975 CHF
ABCAAB3462-100
Anti-alpha 1 Adrenergic Receptor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: alpha 1 Adrenergic Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR