Anti-GNB1 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB3433-100EA 508.36 CHF
ABCAAB3433-100
Anti-GNB1 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: GNB1 (guanine nucleotide binding protein (G protein), beta polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR