Anti-SCAMP5 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3432-100EA 508.36 CHF
ABCAAB3432-100
Anti-SCAMP5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SCAMP5 (secretory carrier membrane protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR