Anti-SCAMP2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3431-100EA 891 CHF
ABCAAB3431-100
Anti-SCAMP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SCAMP2 (secretory carrier membrane protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR