Anti-MRP1 Rat Monoclonal Antibody [clone: MRPr1]

Lieferant: Abcam
ABCAAB3368-250EA 510.34 CHF
ABCAAB3368-250
Anti-MRP1 Rat Monoclonal Antibody [clone: MRPr1]
Antikörper
Type: Primary
Antigen: MRP1
Clonality: monoclonal
Clone: MRPr1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR