Anti-Blood Group A Antigen Mouse monoclonal antibody [clone: T36]

ABCAAB3353-500EA 699 CHF
ABCAAB3353-500
Anti-Blood Group A Antigen Mouse monoclonal antibody [clone: T36]
Type: Primary
Antigen: Blood Group A Antigen
Clonality: monoclonal
Clone: T36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR