Anti-SYNGAP1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3344-100EA 508.36 CHF
ABCAAB3344-100
Anti-SYNGAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SYNGAP1 (synaptic Ras GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR