Anti-Proteasome 20S LMP2 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB3328-100EA 539.48 CHF
ABCAAB3328-100
Anti-Proteasome 20S LMP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Proteasome 20S LMP2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR