Anti-PSMC5 Rabbit polyclonal antibody

ABCAAB3323-100EA 716 CHF
ABCAAB3323-100
Anti-PSMC5 Rabbit polyclonal antibody
Type: Primary
Antigen: PSMC5 (proteasome (prosome, macropain) 26S subunit, ATPase, 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR