Anti-Adenovirus Fiber monomer and trimer Mouse monoclonal antibody [clone: 4D2]

ABCAAB3233-500EA 716 CHF
ABCAAB3233-500
Anti-Adenovirus Fiber monomer and trimer Mouse monoclonal antibody [clone: 4D2]
Type: Primary
Antigen: Adenovirus Fiber monomer and trimer
Clonality: monoclonal
Clone: 4D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR