Anti-MITF Mouse Monoclonal Antibody [clone: D5]

Supplier: Abcam
ABCAAB3201-100EA 466.86 CHF
ABCAAB3201-100
Anti-MITF Mouse Monoclonal Antibody [clone: D5]
Antibodies
Type: Primary
Antigen: MITF (microphthalmia-associated transcription factor)
Clonality: monoclonal
Clone: D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR