Anti-L1CAM Mouse monoclonal antibody [clone: UJ127]

ABCAAB3200-500EA 749 CHF
ABCAAB3200-500
Anti-L1CAM Mouse monoclonal antibody [clone: UJ127]
Type: Primary
Antigen: L1CAM (L1 cell adhesion molecule)
Clonality: monoclonal
Clone: UJ127
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR