Anti-PXN Mouse monoclonal antibody [clone: 5H11]

ABCAAB3127-500EA 741 CHF
ABCAAB3127-500
Anti-PXN Mouse monoclonal antibody [clone: 5H11]
Type: Primary
Antigen: PXN (paxillin)
Clonality: monoclonal
Clone: 5H11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR