Anti-VEGFA Mouse monoclonal antibody [clone: 5C3.F8]

ABCAAB3109-500EA 732 CHF
ABCAAB3109-500
Anti-VEGFA Mouse monoclonal antibody [clone: 5C3.F8]
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: 5C3.F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR