Anti-THY1 Rat Monoclonal Antibody [clone: IBL-6/23]
Lieferant: Abcam
ABCAAB3105-100EA 472.19 CHF
ABCAAB3105-100
Anti-THY1 Rat Monoclonal Antibody [clone: IBL-6/23]
Antikörper
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: IBL-6/23
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse