Anti-ERBB4 Mouse monoclonal antibody [clone: H4.77.16]

ABCAAB3104-500EA 699 CHF
ABCAAB3104-500
Anti-ERBB4 Mouse monoclonal antibody [clone: H4.77.16]
Type: Primary
Antigen: ERBB4 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4)
Clonality: monoclonal
Clone: H4.77.16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR