Anti-COL2 Mouse monoclonal antibody [clone: 5B2.5]

ABCAAB3092-500EA 727 CHF
ABCAAB3092-500
Anti-COL2 Mouse monoclonal antibody [clone: 5B2.5]
Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: monoclonal
Clone: 5B2.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR