Anti-FBN1 Mouse monoclonal antibody [clone: 11C1.3]

ABCAAB3090-500EA 0 CHF
ABCAAB3090-500
Anti-FBN1 Mouse monoclonal antibody [clone: 11C1.3]
Type: Primary
Antigen: FBN1 (fibrillin 1)
Clonality: monoclonal
Clone: 11C1.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR