Anti-DNAJA1 Mouse monoclonal antibody [clone: KA2A5.6]

ABCAAB3089-500EA 0 CHF
ABCAAB3089-500
Anti-DNAJA1 Mouse monoclonal antibody [clone: KA2A5.6]
Type: Primary
Antigen: DNAJA1 (DnaJ (Hsp40) homolog, subfamily A, member 1)
Clonality: monoclonal
Clone: KA2A5.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR