Anti-TOP2A Rabbit polyclonal antibody

ABCAAB2987-500EA 699 CHF
ABCAAB2987-500
Anti-TOP2A Rabbit polyclonal antibody
Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR