Anti-PSMD1 Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB2941-100EA 508.36 CHF
ABCAAB2941-100
Anti-PSMD1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSMD1 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR