Anti-RAB11A Goat Polyclonal Antibody

Supplier: Abcam
ABCAAB2921-100EA 912 CHF
ABCAAB2921-100
Anti-RAB11A Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB11A (RAB11A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR