Anti-ATR Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB2905-100EA 567.58 CHF
ABCAAB2905-100
Anti-ATR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ATR (ataxia telangiectasia and Rad3 related)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR