Anti-beta COP Rabbit polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB2899-100EA 539.48 CHF
ABCAAB2899-100
Anti-beta COP Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: beta COP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR