Anti-DNA Polymerase beta Mouse polyclonal antibody

ABCAAB2856-50EA 660 CHF
ABCAAB2856-50
Anti-DNA Polymerase beta Mouse polyclonal antibody
Type: Primary
Antigen: DNA Polymerase beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR