Anti-pan Endothelin Mouse monoclonal antibody [clone: 3B8C9]

ABCAAB2793-100EA 691 CHF
ABCAAB2793-100
Anti-pan Endothelin Mouse monoclonal antibody [clone: 3B8C9]
Type: Primary
Antigen: pan Endothelin
Clonality: monoclonal
Clone: 3B8C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR