Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]

Supplier: Abcam
ABCAAB2791-100EA 953 CHF
ABCAAB2791-100
Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]
Antibodies
Type: Primary
Antigen: HSP90B1 (heat shock protein 90kDa beta (Grp94), member 1)
Clonality: monoclonal
Clone: 9G10
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR