Anti-HSPB1 Mouse Monoclonal Antibody [clone: G3.1]

Supplier: Abcam
ABCAAB2790-100EA 570.61 CHF
ABCAAB2790-100
Anti-HSPB1 Mouse Monoclonal Antibody [clone: G3.1]
Antibodies
Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: monoclonal
Clone: G3.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR