Anti-HSPA8 Mouse Monoclonal Antibody [clone: 13D3]

Supplier: Abcam
ABCAAB2788-50EA 975 CHF
ABCAAB2788-50
Anti-HSPA8 Mouse Monoclonal Antibody [clone: 13D3]
Antibodies
Type: Primary
Antigen: HSPA8 (heat shock 70kDa protein 8)
Clonality: monoclonal
Clone: 13D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR