Anti-HSPA13 Mouse Monoclonal Antibody [clone: 5A5]

Supplier: Abcam
ABCAAB2787-250EA 570.61 CHF
ABCAAB2787-250
Anti-HSPA13 Mouse Monoclonal Antibody [clone: 5A5]
Antibodies
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: 5A5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR