Anti-Calcium Pump PMCA4 ATPase Mouse Monoclonal Antibody [clone: JA9]

Lieferant: Abcam
ABCAAB2783-100EA 548.5 CHF
ABCAAB2783-100
Anti-Calcium Pump PMCA4 ATPase Mouse Monoclonal Antibody [clone: JA9]
Antikörper
Type: Primary
Antigen: Calcium Pump PMCA4 ATPase
Clonality: monoclonal
Clone: JA9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR