Anti-HIF1 beta Mouse Monoclonal Antibody [clone: 2B10]
Lieferant: Abcam
ABCAAB2771-100EA 567.58 CHF
ABCAAB2771-100
Anti-HIF1 beta Mouse Monoclonal Antibody [clone: 2B10]
Antikörper
Type: Primary
Antigen: HIF1 beta
Clonality: monoclonal
Clone: 2B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now